COM448632937 Combi-Blocks, Inc.

[3-(Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]methanol

  • MDL number MFCD18733401
  • Cas Number : 1809447-76-4
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1651

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1651
Availability details for
Quantity
  • Availability
  • Add to Cart