COM448632947 Combi-Blocks, Inc.

5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3,4-diamine

  • MDL number MFCD27936455
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1661

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1661
Availability details for
Quantity
  • Availability
  • Add to Cart