COM448632952 Combi-Blocks, Inc.

tert-Butyl N-propyl-N-[5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate

  • MDL number MFCD27936459
  • Molecular Weight : 362.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1666

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1666
Availability details for
Quantity
  • Availability
  • Add to Cart