COM448632956 Combi-Blocks, Inc.

2-Chloro-N-methoxy-N-methyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxamide

  • MDL number MFCD27936462
  • Molecular Weight : 326.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1670

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1670
Availability details for
Quantity
  • Availability
  • Add to Cart