COM448632957 Combi-Blocks, Inc.

3-fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-4-carbaldehyde

  • MDL number MFCD18724747
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1671
Availability details for
Quantity
  • Availability
  • Add to Cart