COM448632960 Combi-Blocks, Inc.

6-Hydrazinyl-2-methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD27936464
  • Cas Number : 1353746-95-8
  • Molecular Weight : 249.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1674
Availability details for
Quantity
  • Availability
  • Add to Cart