COM448632963 Combi-Blocks, Inc.

Ethyl 6-methyl-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD27936467
  • Molecular Weight : 330.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1677

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1677
Availability details for
Quantity
  • Availability
  • Add to Cart