COM448632966 Combi-Blocks, Inc.

2-Hydrazinyl-3-nitro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD27936470
  • Cas Number : 1356829-95-2
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1680

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1680
Availability details for
Quantity
  • Availability
  • Add to Cart