COM448632967 Combi-Blocks, Inc.

8-Nitro-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD27936471
  • Molecular Weight : 290.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1681

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1681
Availability details for
Quantity
  • Availability
  • Add to Cart