COM448632968 Combi-Blocks, Inc.

N-Ethyl-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxamide

  • MDL number MFCD27936472
  • Cas Number : 1807629-65-7
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1682

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1682
Availability details for
Quantity
  • Availability
  • Add to Cart