COM448632971 Combi-Blocks, Inc.

N-Butyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide

  • MDL number MFCD27936475
  • Molecular Weight : 304.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1685

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1685
Availability details for
Quantity
  • Availability
  • Add to Cart