COM448632977 Combi-Blocks, Inc.

2-{[3-Nitro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]amino}ethanol

  • MDL number MFCD27936480
  • Cas Number : 1416061-98-7
  • Molecular Weight : 309.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1691

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1691
Availability details for
Quantity
  • Availability
  • Add to Cart