COM448632979 Combi-Blocks, Inc.

N-[(4-Methoxyphenyl)methyl]-3-nitro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

  • MDL number MFCD27936482
  • Molecular Weight : 385.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1693
Availability details for
Quantity
  • Availability
  • Add to Cart