COM448632982 Combi-Blocks, Inc.

N,N-Diethyl-5-nitro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

  • MDL number MFCD27936484
  • Cas Number : 1351395-80-6
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1696

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1696
Availability details for
Quantity
  • Availability
  • Add to Cart