COM448632983 Combi-Blocks, Inc.

5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-thiol

  • MDL number MFCD27936485
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1697

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1697
Availability details for
Quantity
  • Availability
  • Add to Cart