COM448632984 Combi-Blocks, Inc.

N-[5-Amino-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]acetamide

  • MDL number MFCD27936486
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1698

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1698
Availability details for
Quantity
  • Availability
  • Add to Cart