COM448632987 Combi-Blocks, Inc.

4-Hydroxy-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxylic acid

  • MDL number MFCD27936488
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1701

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1701
Availability details for
Quantity
  • Availability
  • Add to Cart