COM448632988 Combi-Blocks, Inc.

3-(Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2,4-diol

  • MDL number MFCD26520015
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1702

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1702
Availability details for
Quantity
  • Availability
  • Add to Cart