COM448632995 Combi-Blocks, Inc.

Ethyl 2-cyano-2-[(2Z)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyridin-2-ylidene]acetate

  • MDL number MFCD27936495
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1709

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1709
Availability details for
Quantity
  • Availability
  • Add to Cart