COM448632996 Combi-Blocks, Inc.

1,3-Dimethyl 2-[5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl]propanedioate

  • MDL number MFCD27936496
  • Molecular Weight : 336.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1710

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1710
Availability details for
Quantity
  • Availability
  • Add to Cart