COM448633000 Combi-Blocks, Inc.

1-(2-Hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone

  • MDL number MFCD18732919
  • Cas Number : 1246560-24-6
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1714

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1714
Availability details for
Quantity
  • Availability
  • Add to Cart