COM448633001 Combi-Blocks, Inc.

4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD27936500
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1715

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1715
Availability details for
Quantity
  • Availability
  • Add to Cart