COM448633002 Combi-Blocks, Inc.

2-[3-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-indole

  • MDL number MFCD27936501
  • Cas Number : 1333340-59-2
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1716

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1716
Availability details for
Quantity
  • Availability
  • Add to Cart