COM448633004 Combi-Blocks, Inc.

4-Methoxycarbonyl-2-trifluoromethylphenylboronic acid pinacol ester

  • MDL number MFCD23379538
  • Cas Number : 1371589-09-1
  • Molecular Weight : 330.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1718

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1718
Availability details for
Quantity
  • Availability
  • Add to Cart