COM448633005 Combi-Blocks, Inc.

Methyl 5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pentanoate

  • MDL number MFCD27936503
  • Cas Number : 1985620-97-0
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1719

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1719
Availability details for
Quantity
  • Availability
  • Add to Cart