COM448633008 Combi-Blocks, Inc.

2-Formyl-4-nitrophenylboronic acid pinacol ester

  • MDL number MFCD22493739
  • Cas Number : 1351054-91-5
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1722

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1722
Availability details for
Quantity
  • Availability
  • Add to Cart