COM448633014 Combi-Blocks, Inc.

4-Acetyl-2-fluorobenzeneboronic acid pinacol ester

  • MDL number MFCD18732910
  • Cas Number : 1807685-51-3
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1728
Availability details for
Quantity
  • Availability
  • Add to Cart