COM448633015 Combi-Blocks, Inc.

2-(3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid

  • MDL number MFCD24368697
  • Cas Number : 1431548-94-5
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1729

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1729
Availability details for
Quantity
  • Availability
  • Add to Cart