COM448633018 Combi-Blocks, Inc.

1-(4-Methylphenyl)-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)ethanone

  • MDL number MFCD27936512
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1733
Availability details for
Quantity
  • Availability
  • Add to Cart