COM448633020 Combi-Blocks, Inc.

6-(Tetramethyl-1,3,2-dioxaborolan-2-yl)hexanoic acid

  • MDL number MFCD27936514
  • Cas Number : 1557159-88-2
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1735

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1735
Availability details for
Quantity
  • Availability
  • Add to Cart