COM448633021 Combi-Blocks, Inc.

2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[4-(trifluoromethyl)phenyl]ethanone

  • MDL number MFCD27936515
  • Molecular Weight : 314.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1736

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1736
Availability details for
Quantity
  • Availability
  • Add to Cart