COM448633025 Combi-Blocks, Inc.

[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indol-3-yl]acetic acid

  • MDL number MFCD27936518
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1740
Availability details for
Quantity
  • Availability
  • Add to Cart