COM448633030 Combi-Blocks, Inc.

Benzoic acid, 4-cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, ethyl ester

  • MDL number MFCD22494140
  • Cas Number : 1126479-87-5
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1745
Availability details for
Quantity
  • Availability
  • Add to Cart