COM448633031 Combi-Blocks, Inc.

2-Formylbenzofuran-5-boronic acid pinacol ester

  • MDL number MFCD25541632
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1746
Availability details for
Quantity
  • Availability
  • Add to Cart