COM448633037 Combi-Blocks, Inc.

7-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-quinazolin-4-one

  • MDL number MFCD27936527
  • Cas Number : 1009303-80-3
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1753

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1753
Availability details for
Quantity
  • Availability
  • Add to Cart