COM448633039 Combi-Blocks, Inc.

tert-Butyl N-[2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazol-4-yl]carbamate

  • MDL number MFCD27936529
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1755
Availability details for
Quantity
  • Availability
  • Add to Cart