COM448633041 Combi-Blocks, Inc.

2-Formyl-4-hydroxy-5-methoxyphenylboronic acid, pinacol ester

  • MDL number MFCD24842538
  • Cas Number : 154750-11-5
  • Molecular Weight : 278.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1759

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1759
Availability details for
Quantity
  • Availability
  • Add to Cart