COM448633047 Combi-Blocks, Inc.

2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)pentanoic acid

  • MDL number MFCD27936536
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1765

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1765
Availability details for
Quantity
  • Availability
  • Add to Cart