COM448633049 Combi-Blocks, Inc.

Ethyl 2-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole-4-carboxylate

  • MDL number MFCD27936538
  • Molecular Weight : 359.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1767

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1767
Availability details for
Quantity
  • Availability
  • Add to Cart