COM448633065 Combi-Blocks, Inc.

N-[5-Methyl-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]acetamide

  • MDL number MFCD27936552
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1785

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1785
Availability details for
Quantity
  • Availability
  • Add to Cart