COM448633068 Combi-Blocks, Inc.

8-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine

  • MDL number MFCD27936554
  • Molecular Weight : 261.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1788

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1788
Availability details for
Quantity
  • Availability
  • Add to Cart