COM448633069 Combi-Blocks, Inc.

6-Fluoro-2-methyl-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)-2,4-dihydro-1,4-benzoxazin-3-one

  • MDL number MFCD27936555
  • Molecular Weight : 307.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1789

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1789
Availability details for
Quantity
  • Availability
  • Add to Cart