COM448633070 Combi-Blocks, Inc.

6-Fluoro-2,2-dimethyl-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one

  • MDL number MFCD27936556
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1790

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1790
Availability details for
Quantity
  • Availability
  • Add to Cart