COM448633075 Combi-Blocks, Inc.

3-fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbaldehyde

  • MDL number MFCD18728070
  • Cas Number : 1417200-18-0
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1795

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1795
Availability details for
Quantity
  • Availability
  • Add to Cart