COM448633078 Combi-Blocks, Inc.

6-Methyl-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD27936563
  • Molecular Weight : 302.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1798

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1798
Availability details for
Quantity
  • Availability
  • Add to Cart