COM448633080 Combi-Blocks, Inc.

N-Methyl-n-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)acetamide

  • MDL number MFCD24386093
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1801

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1801
Availability details for
Quantity
  • Availability
  • Add to Cart