COM448633082 Combi-Blocks, Inc.

3-(Dimethylamino)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbaldehyde

  • MDL number MFCD27936567
  • Cas Number : 1352819-35-2
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1803

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1803
Availability details for
Quantity
  • Availability
  • Add to Cart