COM448633087 Combi-Blocks, Inc.

6-(Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carbaldehyde

  • MDL number MFCD27936572
  • Cas Number : 1381940-44-8
  • Molecular Weight : 282.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1808
Availability details for
Quantity
  • Availability
  • Add to Cart