COM448633093 Combi-Blocks, Inc.

3-Cyclopropyl-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

  • MDL number MFCD27936578
  • Cas Number : 1650548-68-7
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1814

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1814
Availability details for
Quantity
  • Availability
  • Add to Cart