COM448633095 Combi-Blocks, Inc.

1-[2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-1-one

  • MDL number MFCD26390270
  • Cas Number : 1365803-10-6
  • Molecular Weight : 278.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1816

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1816
Availability details for
Quantity
  • Availability
  • Add to Cart